Skip to Content
Merck
All Photos(1)

Key Documents

BMSD113260

Benchmark Zirconium Oxide Bulk Beads

bead diam. 6 mm, Ceria Stabilized

Synonym(s):

grinding medium, grinding beads, beads for homogenization

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

zirconia (oxide)

feature

Ceria Stabilized

packaging

pkg of 50 ea

manufacturer/tradename

Benchmark Scientific D1132-60

bead diam.

6 mm

compatibility

BeadBug and BeadBlaster

General description

Loose, bulk satellites, Ceria Stabilized zirconium, 6mm diameter, TriplePure Molecular Biology Grade for bead homogenization

Application

bead, homogenization, lysis, zirconium, TriplePure, BeadBug, BeadBlaster

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service