Skip to Content
Merck
All Photos(5)

Key Documents

HF07502XSS

Millipore

Hi-Flow Plus Membrane 75

backing thickness 2 mil (50 μm), filter W × L 30.5 cm × 25 cm, pack of 5 sheets

Synonym(s):

HF, Hi-Flow, Hi-Flow Plus HF07502, immunoassay membrane, lateral flow membrane, nitrocellulose membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NA.28

packaging

pack of 5 sheets

Quality Level

parameter

75 sec/4 cm capillary flow rate

backing thickness

2 mil (50 μm)

filter W × L

30.5 cm × 25 cm

General description

Hi-Flow Plus membranes are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. Lateral flow tests faciliate quick and convenient detection / quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine. Hi-Flow Plus membranes are ideally suited for such applications. Available in a variety of capillary flow times and backing options, choosing Hi-Flow Plus membranes can improve assay performance and offer flexibility to meet a wide range of assay design requirements.

Hi-Flow Plus 75 (HF075) is suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern. This Hi-Flow Plus 75 pack contains 5 sample sheets of 2 mil backed nitrocellulose membrane with a nominal capillary flow time of 75 seconds/4 cm.

Application

  • Hi-Flow Plus membrane is a critical raw material suitable for use in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs, etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.
  • It is suitable for use with all lateral flow test techniques including competitive, sandwich, and multiplex detection assays.
  • It is optimised for use in both qualitative and quantitative test types and pairs well with lateral flow detector particles, including colloidal gold nanoparticles, dyed/ fluorescent microspheres, etc.
  • It is suitable for use in lateral flow tests designed to detect analytes present in a broad range of samples matrices including, blood, saliva, urine, nasopharyngeal swabs, sweat, and others.

Features and Benefits

FEATURES:

  • Consistent flow and surface chemistry, robust manufacturing process.
  • Proprietary surface chemistry optimised for lateral flow applications.
  • High Quality Level: MQ400.
  • Trusted partner and brand with decades of membrane manufacturing experience.
  • High production capacity, high stock levels, multiple manufacturing lines.
  • Convenient samples packs available in several formats.
  • Manufacturing scale quantities available to purchase online with immediate shipping.
  • We offer a complete solution; along with Hi-Flow Plus membranes we offer raw materials across the entire lateral flow workflow as well as assay development services: https://www.sigmaaldrich.com/US/en/services/ivd-development-manufacturing/lateral-flow-services.
BENEFITS:

  • Consistent capillary flow time and membrane thickness.
  • Superior surface morphology allows for consistent and even flow distribution of reagents across the membrane surface.
  • Sharp, intense, and uniform test and control line formation.
  • Excellent intra-lot and inter-lot consistency.
  • Better assay sensitivity.
  • Excellent reproducibility.
  • Improved lateral flow test manufacturing yields.
  • Quick delivery and lead time.
  • No supply disruption.
  • Excellent technical support.

Legal Information

Hi-Flow is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service