Skip to Content
Merck
All Photos(5)

Key Documents

CFSP173000

Millipore

Cellulose Absorbent Sample Pads

thickness 0.83 mm, filter L × W 1.7 cm × 30 cm, pack of 100 strips

Synonym(s):

Absorbent pad, C083 Cellulose Fiber Sample Pad Strips, Cellulose pad, Sample pad

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.23

packaging

pack of 100 strips

Quality Level

manufacturer/tradename

SureWick®

W

17 mm

filter L × W

1.7 cm × 30 cm

thickness

0.83 mm

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

SureWick® Pad Materials are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. SureWick® cellulose fiber pad materials are used as sample and absorbant pads in lateral flow assays. This pack contains 100 strips of 17mm x 30cm C083 cellulose fiber pad material.

Application

SureWick® Pad Materials are optimised for use as sample, conjugate, or absorbent pads in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.

Features and Benefits

  • SureWick® cellulose fiber pads are made from non-woven, 100% pure cellulose fiber.
  • These pads do not contain binders or glues, which can interfere with assay performance.
  • Manufactured to ensure excellent consistency, they provide reliable, predictable sample flow.
  • They exhibit consistent thickness, bed volume, and uniform surface quality.
  • Quick delivery and lead time.
  • Backed by excellent technical support.

Legal Information

SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maryam Zafar et al.
Applied microbiology and biotechnology, 104(8), 3391-3402 (2020-02-24)
Birnaviridae is a family of viruses (birnaviruses) which consists of four genera, members of which cause diseases in fish, birds, mollusks, and insects. The genome of birnaviruses encodes the highly immunogenic VP2 capsid protein. In order to demonstrate that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service