Skip to Content
Merck
All Photos(1)

Documents

NRK-52E

87012902, rat kidney, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NRK-52E, 87012902

biological source

rat kidney

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat kidney epithelial

Cell Line Description

Cloned from the same mixed culture of normal rat kidney cells as NRK-49F, but has epithelial-like morphology. It has distinct growth and transforming characteristics distinct from NRK-49F.

Application

NRK-52E has been used to study the effects of altered selenium supply on the expression of endogenous thioredoxin reductase (TrxR).

Culture Medium

DMEM + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 5-10% Foetal Bovine Serum (FBS) or New Born Calf Serum (NBCS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Remove all traces of trypsin from cultures during sub-culturing i.e. spin cells or neutralise with serum.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service