Skip to Content
Merck
All Photos(1)

Documents

NRK-49F

86101301, rat kidney, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NRK-49F, 86101301

biological source

rat kidney

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat kidney fibroblast

Cell Line Description

Derived from normal kidney and exhibits contact-inhibition. It is sensitive to viral or chemical transformation, including proteins such as SGF. Originated from the same mixed culture as NRK-52E but has distinct characteristics.

Application

Contact inhibition and transformation studies

Culture Medium

DMEM + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 5-10% Foetal Bovine Serum (FBS) or New Born Calf Serum (NBCS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. The cells should be maintained sub-confluent, otherwise they will transform.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service