327743
Polystyrene
analytical standard, for GPC, 20,000
Synonym(s):
PS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[CH2CH(C6H5)]n
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
analytical standard
for GPC
Quality Level
form
powder
mol wt
average Mw 20,000 (Typical)
technique(s)
gel permeation chromatography (GPC): suitable
format
neat
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Polystyrene (for GPC, 20,000) is a synthetic thermoplastic, that finds a variety of applications owing to its characteristics like low cost, rigidity, low specific weight, high chemical resistance, mechanical flexibility, biocompatibility and good processability.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Weathering of Polymers (1983)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service