Skip to Content
Merck
Polystyrene analytical standard, for GPC, 300,000

Polystyrene

Polystyrene analytical standard, for GPC, 300,000
Synonym(s):
PS
Linear Formula:
[CH2CH(C6H5)]n

Products

Brand
Color
Manufacturer
Physical Form
Format
Length (m)
Quality Segment
Special Grade
Technique
Showing 1-20 of 50
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3