Skip to Content
Merck
All Photos(1)

Key Documents

00410

Sigma-Aldrich

Ethyl acetoacetate

puriss. p.a., ≥99.0% (GC)

Synonym(s):

Acetoacetic ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOC2H5
CAS Number:
Molecular Weight:
130.14
Beilstein:
385838
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39022303
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.48 (vs air)

Quality Level

vapor pressure

1 mmHg ( 28.5 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

autoignition temp.

580 °F

expl. lim.

9.5 %

impurities

≤0.5% water

refractive index

n20/D 1.419

bp

181 °C (lit.)

mp

−43 °C (lit.)

solubility

water: soluble 130 g/L at 20 °C

density

1.029 g/mL at 20 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.5 mg/kg

functional group

ester

SMILES string

CCOC(=O)CC(C)=O

InChI

1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3

InChI key

XYIBRDXRRQCHLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl acetoacetate can be used as a reactant in the synthesis of:
  • Knoevenagel condensation products by reacting with α-alkylideneacetoacetate and aliphatic, aromatic, heteroaromatic aldehydes.
  • Michael adducts, via Michael addition reaction with chalcones and azachalcones in the presence of a base catalyst.
  • Substituted nicotinic acid derivatives by treating with α, β-unsaturated oximes via Michael addition followed by ring closure reaction.

It can be also utilized as a reactant in the transesterification and asymmetric hydrogenation reactions to produce valuable products.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

164.3 °F - closed cup

Flash Point(C)

73.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael addition of ethyl acetoacetate to α,β -unsaturated oximes in the presence of FeCl3: a novel synthetic route to substituted nicotinic acid derivatives
Chibiryaev AM, et al.
Tetrahedron Letters, 41(41), 8011-8013 (2000)
Knoevenagel condensation reaction between benzaldehyde and ethyl acetoacetate in microreactor and membrane microreactor
Lau WN, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 115(1-2), 156-163 (2008)
Mechanochemical Michael reactions of chalcones and azachalcones with ethyl acetoacetate catalyzed by K2CO3 under solvent-free conditions
Zhang Ze, et al.
Chemistry Letters (Jpn), 33(2), 168-169 (2004)
Enantioselective catalytic asymmetric hydrogenation of ethyl acetoacetate in room temperature ionic liquids
Berthod M, et al.
Tetrahedron Asymmetry, 15(14), 2219-2221 (2004)
Boric acid: an efficient and environmentally benign catalyst for transesterification of ethyl acetoacetate
Kondaiah GCM, et al.
Tetrahedron Letters, 49(1), 106-109 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service