Skip to Content
Merck
Ethyl acetoacetate analytical standard

Ethyl acetoacetate

Ethyl acetoacetate analytical standard
Synonym(s):
Acetoacetic ester, Ethyl acetoacetate
Linear Formula:
CH3COCH2COOC2H5
CAS Number:
Molecular Weight:
130.14
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique