Skip to Content
Merck
All Photos(1)

Key Documents

MABE145

Sigma-Aldrich

Anti-Runx3 Antibody, clone R3-5G4

clone R3-5G4, from mouse

Synonym(s):

Runt-related transcription factor 3, Acute myeloid leukemia 2 protein, Core-binding factor subunit alpha-3, CBF-alpha-3, Oncogene AML-2, Polyomavirus enhancer-binding protein 2 alpha C subunit, PEA2-alpha C, PEBP2-alpha C, SL3-3 enhancer factor 1 alpha C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
R3-5G4, monoclonal
application:
IP
WB
species reactivity:
human
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

R3-5G4, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RUNX3(864)

General description

Runx3 is a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5′-PYGPYGGT-3′ found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer.

Immunogen

Recombinant protein corresponding to human Runx3.

Application

Immunoprecipitation Analysis: A representative lot from an independent laboratory detected Runx3 in HCT116, SW620, and SW480 nuclear extracts. (Ito, K., et al. (2008). Cancer Cell. 14(3):226-237.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-Runx3 Antibody, clone R3-5G4 (mouse monoclonal antibody) validated in WB, IP to detect Runx3 also known as Runt-related transcription factor 3, Acute myeloid leukemia 2 protein, PEA2-alpha C, PEBP2-alpha C.

Quality

Evaluated by Western Blot in Daudi cell lysate.

Western Blot Analysis: 0.25 mg/mL of this antibody detected Runx3 in 10 µg Daudi cell lysate.

Target description

~44 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Daudi cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon I Dreher et al.
Frontiers in cell and developmental biology, 8, 81-81 (2020-03-21)
Guiding progenitor cell development between chondral versus endochondral pathways is still an unachieved task of cartilage neogenesis, and human mesenchymal progenitor cell (MPC) chondrogenesis is considered as a valuable model to better understand hypertrophic development of chondrocytes. Transcription factors Runx2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service