Skip to Content
Merck
All Photos(1)

Key Documents

5.00583

Sigma-Aldrich

(+)-Brefeldin A, Eupenicillium brefeldianum

InSolution, ≥95%, 25 mM in DMSO

Synonym(s):

InSolution (+)-Brefeldin A, Eupenicillium brefeldianum, 1,6,7,8,9,11a,12,13,14,14a-Decahydro-1,13-dihydroxy-6-methyl-4H-cyclopent[f]oxacyclotridecin-4-one, (1 S,2 E,7 S,10 E,12 R,13 R,15 S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one,

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O4
CAS Number:
Molecular Weight:
280.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

O1[C@H](CCC\C=C\[C@H]2[C@H]([C@H](\C=C\C1=O)O)C[C@H](C2)O)C

InChI

1S/C16H24O4/c1-11-5-3-2-4-6-12-9-13(17)10-14(12)15(18)7-8-16(19)20-11/h4,6-8,11-15,17-18H,2-3,5,9-10H2,1H3/b6-4+,8-7+/t11-,12+,13-,14+,15-/m0/s1

InChI key

KQNZDYYTLMIZCT-PNFJWZTBSA-N

General description

Specifically and reversibly blocks translocation of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus without affecting endocytosis or lysosome function. Inhibitor of HDL-mediated cholesterol efflux. Causes disassembly of the Golgi complex and ER swelling in a variety of mammalian cell lines at <40 ng/ml. Inhibits the activity of BIG1 and BIG2, the guanine-nucleotide exchange proteins for ADP-ribosylation factors. Blocks binding of ADP-ribosylation factor to the Golgi and inhibits the GDP-GTP exchange.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ADP-ribosylation factor (ARF)
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C. Use only fresh DMSO for further dilutions prior to use.

Other Notes

Yamagi, R., et al. 2000. Proc. Natl. Acad. Sci. USA97, 2567.
Moriango, N., et al. 1996. Proc. Natl. Acad. Sci. USA93, 12856.
Mendez, A.J. 1995. J. Biol. Chem.270, 5891.
Ridgway, N.D., et al. 1995. J. Biol. Chem. 270, 8023.
Tsai, S.C., et al. 1994. Proc. Natl. Acad. Sci. USA91, 3063.
Ktistakis, N.Y., et al. 1992. Nature356, 344.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service