Skip to Content
Merck
All Photos(1)

Key Documents

880343P

Avanti

PChemsPC

Avanti Research - A Croda Brand 880343P, powder

Synonym(s):

1-palmitoyl-2-cholesterylhemisuccinoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H98NO10P
CAS Number:
Molecular Weight:
964.34
UNSPSC Code:
51191904
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (880343P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880343P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OCC[N+](C)(C)C)OC[C@H](OC(CCC(OC1CC[C@]2(C)C(C1)=CCC3C2CC[C@]4(C)C3CC[C@H]4[C@@H](C)CCCC(C)C)=O)=O)COC(CCCCCCCCCCCCCCC)=O

InChI key

QFPQHJPAWCNJAS-GYPLUMKTSA-N

Application

PChemsPC or 1-palmitoyl-2-cholesterylhemisuccinoyl-sn-glycero-3-phosphocholine might be used to study its pharmacokinetics and biodistribution patterns in comparison with phospholipid-cholesterol liposomes. It might also be used as a component in the monolayer to study its characterization at the air/water interface.

Biochem/physiol Actions

PChemsPC or 1-palmitoyl-2-cholesterylhemisuccinoyl-sn-glycero-3-phosphocholine is a sterol-phospholipid hybrid. Cholesterol incorporation with phospholipids reduces the permeability of the lipid bilayer, organizes phospholipid chains and avoids the phospholipid phase transition. This improves the stability of the vesicles.

Packaging

5 mL Amber Glass Screw Cap Vial (880343P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michał Flasiński et al.
Langmuir : the ACS journal of surfaces and colloids, 32(16), 4095-4102 (2016-04-06)
The two synthetic sterol-phospholipid hybrids DCholPC and PCholPC were investigated in monolayers at the air/water interface. Study was based on π-A isotherm analysis complemented with application of grazing incidence X-ray diffraction. It was found that both compounds are capable of
Aditya G Kohli et al.
Journal of controlled release : official journal of the Controlled Release Society, 176, 86-93 (2013-12-26)
We introduce a method for tracking the rate and extent of delivery of liposome contents in vivo based on encapsulation of 4-methylumbelliferyl phosphate (MU-P), a profluorophore of 4-methylumbelliferone (MU). MU-P is rapidly dephosphorylated by endogenous phosphatases in vivo to form
Maryam Iman et al.
International journal of pharmaceutics, 408(1-2), 163-172 (2011-02-01)
1,2-Di-stigma-steryl-hemi-succinoyl-sn-glycero-3-phosphocholine (DSHemsPC) is a new lipid in which two molecules of stigmasterol (an inexpensive plant sterol) are covalently linked via a succinic acid to glycerophosphocholine. Since amphotericin B (AmB) interacts with sterols, we postulated that DSHemsPC could be used in
Zhaohua Huang et al.
Angewandte Chemie (International ed. in English), 48(23), 4146-4149 (2009-05-09)
Extreme makeover of cholesterol: Cholesterol exchange is a major reason for the instability of liposomes in blood. The formation of a covalent hybrid between cholesterol and glycerophosphocholine preserves the bilayer-stabilizing effect of free cholesterol but prevents its transfer from the
F Foglia et al.
Langmuir : the ACS journal of surfaces and colloids, 27(13), 8275-8281 (2011-06-04)
Langmuir isotherm, neutron reflectivity, and small angle neutron scattering studies have been conducted to characterize the monolayers and vesicular bilayers formed by a novel chimeric phospholipid, ChemPPC, that incorporates a cholesteryl moeity and a C-16 aliphatic chain, each covalently linked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service