I27503
2-Isopropylbenzenethiol
technical grade, 90%
Synonym(s):
2-Isopropylthiophenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2CHC6H4SH
CAS Number:
Molecular Weight:
152.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Assay
90%
refractive index
n20/D 1.5579 (lit.)
bp
220 °C (lit.)
density
1.005 g/mL at 25 °C (lit.)
SMILES string
CC(C)c1ccccc1S
InChI
1S/C9H12S/c1-7(2)8-5-3-4-6-9(8)10/h3-7,10H,1-2H3
InChI key
QEDRUXIMTJVXFL-UHFFFAOYSA-N
Gene Information
human ... GABRA1(2554)
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
186.8 °F - closed cup
Flash Point(C)
86 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service