SRP3203
IFN-lambda 2 from mouse
recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(s):
IL-28A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
mol wt
19.8 kDa
packaging
pkg of 20 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... IL28A(338374)
Biochem/physiol Actions
IFN-λ 2 (Ifnl2) belongs to the type III interferon (IFN-λ) family of proteins. It is associated with antiviral, antitumor and immune activities. In a retinal autoimmunity mouse model, Ifnl2 increases the autoimmune disease. Ifnl2 signals via IFN (interferon)-λ receptor complex consisting of IL-10Rβ (interleukin 10 receptor β) and the IL-28Rα chains. The signaling pathway involves JAK1 (Janus kinase 1)/TYK2 (non-receptor tyrosine-protein kinase) tyrosine kinases, transcription factor complex IFN-stimulated gene factor 3 containing STAT1 (signal transducer and activator of transcription 1), STAT2 and IFN regulatory factor (IRF)-9.
Recombinant murine IFN-l2 is a 19.8kDa protein containing 175 amino acid residues.
Recombinant murine IFN-l2 is a 19.8kDa protein containing 175 amino acid residues.
Sequence
MDPVPRATRL PVEAKDCHIA QFKSLSPKEL QAFKKAKDAI EKRLLEKDMR CSSHLISRAW DLKQLQVQER PKALQAEVAL TLKVWENMTD SALATILGQP LHTLSHIHSQ LQTCTQLQAT AEPKPPSRRL SRWLHRLQEA QSKETPGCLE DSVTSNLFRL LTRDLKCVAS GDQCV
Physical form
Lyophilized from 20 mM Sodium Phosphate, pH 7.5.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
An important role for type III interferon (IFN-lambda/IL-28) in TLR-induced antiviral activity.
Journal of Immunology, 180, 2474-2474 (2008)
Characterization of the mouse IFN-lambda ligand-receptor system: IFN-lambdas exhibit antitumor activity against B16 melanoma.
Cancer Research, 66, 4468-4468 (2006)
Interleukin-28A enhances autoimmune disease in a retinal autoimmunity model.
Cytokine, 70, 179-179 (2014)
FEBS letters, 504(1-2), 36-40 (2001-08-28)
Pregnancy-associated plasma protein-A (PAPP-A) has recently been identified as the proteinase responsible for cleavage of insulin-like growth factor binding protein (IGFBP)-4, an inhibitor of IGF action, in several biological fluids. Cleavage of IGFBP-4 by PAPP-A is believed to occur only
Nature immunology, 4(1), 69-77 (2002-12-17)
We report here the identification of a ligand-receptor system that, upon engagement, leads to the establishment of an antiviral state. Three closely positioned genes on human chromosome 19 encode distinct but paralogous proteins, which we designate interferon-lambda1 (IFN-lambda1), IFN-lambda2 and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service