Skip to Content
Merck
All Photos(1)

Documents

SRP3059

Sigma-Aldrich

Interferon-λ human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

IFN-λ human, IL-29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.2-0.5 ng/mL ED50

mol wt

19.8 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL29(282618)

General description

IFN-λ-1,-2, and -3 (also known as IL-29, IL-28A and IL-28B respectively) are distantly related to the IL-10 family of interferons. All three IFN-λs use a distinct receptor system composed of an IFN-λR1 subunit (also called CRF2-12) and IL-10R2 subunit (also called CRF2-14). Signaling through this receptor system induces antiviral defense similar but distinct than the type I Interferons. Recombinant human IFN-L1 is a 19.8kDa protein containing 178 amino acid residues.

Biochem/physiol Actions

Interleukin-29 (IL-29) enhances the production of interleukins from monocytes. It has been observed that it stimulates antiviral responses to herpes simplex virus 1 in vitro. Serums from patients with rheumatoid arthritis have high levels of IL29.

Sequence

PTSKPTTTGK GCHIGRFKSL SPQELASFKK ARDALEESLK LKNWSCSSPV FPGNWDLRLL QVRERPVALE AELALTLKVL EAAAGPALED VLDQPLHTLH HILSQLQACI QPQPTAGPRP RGRLHHWLHR LQEAPKKESA GCLEASVTFN LFRLLTRDLK YVADGNLCLR TSTHPEST

Physical form

Lyophilized from 1x PBS.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of human immune interferon cDNA in E. coli and monkey cells.
P W Gray et al.
Nature, 295(5849), 503-508 (1982-02-11)
Vivienne A Ezzat et al.
Diabetes, obesity & metabolism, 10(3), 198-211 (2008-02-14)
Patients with insulin resistance and type 2 diabetes have an excessive risk of cardiovascular disease (CVD); this increased risk is not fully explained by traditional risk factors such as hypertension and dyslipidaemias. There is now compelling evidence to suggest that
Qian Wu et al.
Autoimmunity, 46(1), 40-43 (2012-10-09)
Interferon (IFN)-λ1 is a newly described cytokine that is known for its proinflammatory activity in viral infection and in cancer. Because recent studies indicated that IFN-λ can influence significantly the innate and adaptive immune response, we studied IFN-λ in a
K Lopušná et al.
Acta virologica, 58(4), 325-332 (2014-12-19)
Lambda interferons (IFN-λ) are known to induce potent antiviral response in a wide variety of target cells. They activate the same intracellular signalling pathways and have similar biological activities as IFN-α/β, including antiviral activity, but signal via distinct receptor complex
Fang Wang et al.
Arthritis research & therapy, 14(5), R228-R228 (2012-10-20)
The immunoregulatory function of interleukin (IL)-29 has recently been recognized. However, little is known about the involvement of IL-29 in the pathogenesis of rheumatoid arthritis (RA). This study aimed to examine the expression profiles of IL-29 in blood, synovial fluid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service