Skip to Content
Merck
All Photos(1)

Key Documents

SML3569

Sigma-Aldrich

M4K2234

≥98% (HPLC)

Synonym(s):

2-Fluoro-4-(5-(4-(4-isopropylpiperazin-1-yl)phenyl)-4-methylpyridin-3-yl)-6-methoxybenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31FN4O2
CAS Number:
Molecular Weight:
462.56
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C)N(CC1)CCN1C2=CC=C(C3=C(C)C(Cl)=CN=C3)C=C2

InChI

1S/C19H24ClN3/c1-14(2)22-8-10-23(11-9-22)17-6-4-16(5-7-17)18-12-21-13-19(20)15(18)3/h4-7,12-14H,8-11H2,1-3H3

InChI key

UFAWMCANSWTKEG-UHFFFAOYSA-N

Biochem/physiol Actions

M4K2234 is a potent and selective TGF-β type I receptors ALK1/ACVRL1 & ALK2/ACVR1 inhibitor (IC50 = 7/14 nM, [ATP] = 10 mM) that inhibits TNIK and ALK6/BMPR1B only at higher concentrations (IC50 = 41 & 88 nM, respectively) with good kinome selectivity over >370 other kinases, including ALK3/BMPR1A, ALK4/ACVR1B, ALK5/TGFBR1 (IC50 = 0.165, 1.66, 1.95 μM, respectively). M4K2234 inhibits cellular phospho-SMAD1/5/8 induction by 10 ng/mL BMP-7 and phospho-SMAD2 induction by either 10 ng/mL activin A or 100 ng/mL TGF-β1 with good target selectivity by cellular NanoBRET assays (ALK1/2 IC50 = 83/13 nM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deeba Ensan et al.
Journal of medicinal chemistry, 63(9), 4978-4996 (2020-05-06)
Diffuse intrinsic pontine glioma is an aggressive pediatric cancer for which no effective chemotherapeutic drugs exist. Analysis of the genomic landscape of this disease has led to the identification of the serine/threonine kinase ALK2 as a potential target for therapeutic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service