CLS3356
Corning® 96 Well Storage Microplate
round bottom, black polypropylene, bag of 25 ×, non-sterile, lid: no
Synonym(s):
Storage Microplate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material:
black bottom
black polypropylene
round bottom
black polypropylene
round bottom
size:
96 wells
well volume:
360 μL
well working volume:
75-200 μL
Recommended Products
material
black bottom
black polypropylene
round bottom
sterility
non-sterile
feature
lid: no
manufacturer/tradename
Corning 3356
packaging
bag of 25 ×
size
96 wells
well volume
360 μL
well working volume
75-200 μL
color
black
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Corning® 96 well microplates support various assay and storage application requirements.
- Features uniform skirt heights for the greater robotic gripping surface.
- Resistant to most solvents including DMSO, ethanol, methanol.
- RNase-/DNase-free.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service