Skip to Content
Merck
All Photos(1)

Key Documents

C-12909

Sigma-Aldrich

Human CD14+ Monocytes (hMoCD14+-PB)

Isolated from peripheral blood, single donor, 10 million cryopreserved cells

Synonym(s):

hMoCD14+-PB cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

Pricing and availability is not currently available.

biological source

human peripheral blood

packaging

pkg of 10,000,000 cells

morphology

(monocytes)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Blood

Application

Monocytes (Mo) are immature phagocytic cells circulating in the blood. Acting as antigen-presenting immune cells, they can phagocytize and degrade microbes and particulate matter. Monocytes can differentiate into several different cell types. The most common applications for Monocytes are the in vitro differentiation into monocyte-derived dendritic cells (moDC), macrophages, and osteoclasts as well as their use as phagocytic immune cells in immunology and infection biology.CD14 belongs to the family of LPS receptor antigens and is strongly expressed on the majority of Monocytes. Thus, binding of immunomagnetic particles to this antigen is employed for purification of PromoCell CD14+-Monocytes (hMoCD14+-PB) from Mononuclear Cells (MNC). Human CD14+ Monocytes are isolated from fresh peripheral blood. After isolation of the ultra-pure mononuclear cell fraction by proprietary methods, the CD14+ Monocytes are isolated by positive selection with >95% purity.

Quality

Rigid quality control tests are performed for each lot of Monocytes. They are routinely characterized by flow cytometry analyzing the viability, cell size and purity. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 1 x 106 cells per mL

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore over 350 PromoCell products on Sigma.com, including cryopreserved blood cells and cell culture media for various human cells.

Explore over 350 PromoCell products on Sigma.com, including cryopreserved blood cells and cell culture media for various human cells.

Explore over 350 PromoCell products on Sigma.com, including cryopreserved blood cells and cell culture media for various human cells.

Explore over 350 PromoCell products on Sigma.com, including cryopreserved blood cells and cell culture media for various human cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service