Skip to Content
Merck
All Photos(1)

Key Documents

34888-M

Sigma-Aldrich

Acetonitrile

≥99.9%, for HPLC, for UV, suitable for HPLC

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
41116004
NACRES:
NA.03
grade:
for HPLC, for UV
Assay:
≥99.9%
technique(s):
HPLC: suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)

Product Name

Acetonitrile, for HPLC, for UV, ≥99.9%

grade

for HPLC, for UV

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

HPLC: suitable

impurities

≤0.0002% free alkali (NH3)
≤0.002% free acid (as CH3COOH)
≤0.02% water
≤0.5 ppb fluorescence (quinine) at 365 nm
≤1 ppb fluorescence (quinine) at 254 nm

evapn. residue

≤0.0005%

transmittance

195 nm, ≥65%
200 nm, ≥88%
210 nm, ≥93%
235 nm, ≥99%

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile is a widely used organic solvent in reversed-phase liquid chromatography due to its high miscibility with water, suitable UV absorbance properties, and low viscosity characteristics.

Application

Acetonitrile can be used as a component in:
  • Mobile phase mixture for the HPLC analysis, to determine the purity of ester content (%FAME).
  • Mobile phase for the ultra-performance liquid chromatography (UPLC) analysis of perfluoroalkyl carboxylic and sulfonic compounds.
  • Mobile phase for high-performance liquid chromatography (HPLC), enabling the effective separation of TMP (trimethoprim) and SMX (sulfamethoxazole).
  • For sample preparation, including extraction, dilution, and dissolutionof compounds prior to analysis.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic model for sonolytic degradation of non-volatile surfactants: Perfluoroalkyl substances
Shende T, et al.
Ultrasonics Sonochemistry, 51, 359-368 (2019)
A rapid, acetonitrile-free, HPLC method for determination of melamine in infant formula
Venkatasami G, et al.
Analytica Chimica Acta, 665(2), 227-230 (2010)
Effects of the antibiotics trimethoprim (TMP) and sulfamethoxazole (SMX) on granulation, microbiology, and performance of aerobic granular sludge systems
Barros ARM, et al.
Chemosphere, 262, 127840-127840 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service