Skip to Content
Merck
All Photos(2)

Key Documents

4.80672

Supelco

Acetonitrile solution

suitable for HPLC, LiChrosolv®, contains 0.05% (v/v) trifluoroacetic acid

Synonym(s):

Acetonitrile solution, ACN, ACN + TFA, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
4124158
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-835-2
NACRES:
NA.03
technique(s):
HPLC: suitable
bp:
81.6 °C/1,013 hPa
vapor pressure:
97 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Acetonitrile with 0.05% (v:v) Trifluoroacetic acid, LiChrosolv®

vapor pressure

97 hPa ( 20 °C)

Quality Level

description

isocratic

product line

LiChrosolv®

form

liquid

concentration

0.045-0.055 % (v/v) (trifluoroacetic acid)

technique(s)

HPLC: suitable

bp

81.6 °C/1,013 hPa

transition temp

flash point 2 °C (Information refers to the main component.)

density

0.782 g/mL at 25 °C

format

mixture

storage temp.

2-30°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Analysis Note

Assay (trifluoroacetic acid): 0.045 - 0.055 % (v/v)

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - Information refers to the main ingredient.

Flash Point(C)

2 °C - Information refers to the main ingredient.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service