Skip to Content
Merck
All Photos(1)

Documents

ZWBASE012

Milli-Q

ROCare

Base cleaning chemical for removing organics, grime and slime from RO membranes

Synonym(s):

ROCare B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104210
eCl@ss:
33050190
NACRES:
JA.14

Quality Level

feature

removes organics, grime and slime from RO membranes

packaging

pkg of 12 packs

storage condition

dry at room temperature (Tightly closed)

compatibility

for use with Elix® Advantage
for use with Milli-Q® HR
for use with Milli-Q® HX
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

ROCare B is a lab water purification consumable that ensures the system′s lifetime and performance by effectively removing some or most organic material buildup on the surface of the reverse osmosis (RO) membranes.
The base cleaner is delivered in a pouch and the empty pouch is removed after use. It is periodically used when prompted by the system or when the RO % rejection drops more than 5% or when the RO permeate flow rate drops more than 10% due to organic material build up.

Application

ROCare B base cleaning pouches protect RO membranes that support Elix® Advantage, Milli-Q® HR 7000, Milli-Q® HX 7000 and Milli-Q® IX 7003/05/10/15 water purification systems.

Features and Benefits

  • Base cleaning chemical for removing organics, grime and slime
  • pH 10 in an aqueous solution
  • Contains 12 packages

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Respiratory system, Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service