Skip to Content
Merck
All Photos(2)

Key Documents

MABE1803

Sigma-Aldrich

Anti-SPT5 Antibody, clone 6F1

clone 6F1, from rat

Synonym(s):

Transcription elongation factor SPT5, hSPT5, DRB sensitivity-inducing factor 160 kDa subunit, DSIF p160, DRB sensitivity-inducing factor large subunit, DSIF large subunit, Tat-co-transactivator 1 protein, Tat-CT1 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F1, monoclonal

species reactivity

human

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SUPT5H(6829)

General description

Transcription elongation factor SPT5 (UniProt: O00267; also known as hSPT5, DRB sensitivity-inducing factor 160 kDa subunit, DSIF p160, DRB sensitivity-inducing factor large subunit, DSIF large subunit, Tat-co-transactivator 1 protein, Tat-CT1 protein) is encoded by the SUPT5H (also known as SPT5, SPT5H) gene (Gene ID: 6829) in human. SPT5 is a component of the DRB sensitivity-inducing factor complex (DSIF complex), which regulates mRNA processing and transcription elongation by RNA polymerase II. DSIF is a heterodimer of SPT4 and SPT5 that positively regulates mRNA capping by stimulating the mRNA guanylyltransferase activity of RNGTT/CAP1A. DSIF also acts cooperatively with the negative elongation factor complex (NELF complex) to enhance transcriptional pausing at sites proximal to the promoter. Transcriptional pausing can facilitate the assembly of an elongation competent RNA polymerase II complex. DSIF and NELF promote pausing by inhibition of the transcription elongation factor TFIIS/S-II, which binds to RNA polymerase II at transcription pause sites and stimulates the weak intrinsic nuclease activity of the enzyme. Cleavage of blocked transcripts by RNA polymerase II promotes the resumption of transcription from the new 3′ terminus and may allow repeated attempts at transcription through natural pause sites. DSIF can also positively regulate transcriptional elongation and is required for the efficient activation of transcriptional elongation by the HIV-1 nuclear transcriptional activator, Tat. SPT5 can be methylated by PRMT1/HRMT1L2 and PRMT5/SKB1 and this methylation negatively regulates interaction with the positive transcription elongation factor (P-TEFb) and RNA polymerase II.

Specificity

Clone 6F1 specifically detects SPT5 in HEK293T and Jurkat cells. It targets an epitope of 18 amino acids within the C-terminal half.

Immunogen

Ovallbumin-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human Transcription elongation factor SPT5.

Application

Anti-SPT5, clone 6F1, Cat. No. MABE1803, is a highly specific rat monoclonal antibody that targets Transcription elongation factor SPT5 and has been tested in Chromatin Immunoprecipitation (ChIP) and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected SPT5 in 10 µg of HEK293T cell lysate.

Chromatin Immunoprecipitation Analysis: A representative lot detected SPT5 in Chromatin Immunoprecipitation applications (Kelso, T.W., et. al. (2014). Mol Cell Biol. 34(19):3675-88).

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected SPT5 in 10 µg of Jurkat cell lysate.

Target description

~121 kDa observed; 121 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danya J Martell et al.
Developmental cell, 58(20), 2112-2127 (2023-08-17)
Controlled release of promoter-proximal paused RNA polymerase II (RNA Pol II) is crucial for gene regulation. However, studying RNA Pol II pausing is challenging, as pause-release factors are almost all essential. In this study, we identified heterozygous loss-of-function mutations in
Felipe Beckedorff et al.
Cell reports, 32(3), 107917-107917 (2020-07-23)
Transcription by RNA polymerase II (RNAPII) is pervasive in the human genome. However, the mechanisms controlling transcription at promoters and enhancers remain enigmatic. Here, we demonstrate that Integrator subunit 11 (INTS11), the catalytic subunit of the Integrator complex, regulates transcription
Kaiwei Liang et al.
Cell, 175(3), 766-779 (2018-10-20)
The super elongation complex (SEC) is required for robust and productive transcription through release of RNA polymerase II (Pol II) with its P-TEFb module and promoting transcriptional processivity with its ELL2 subunit. Malfunction of SEC contributes to multiple human diseases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service