Merck
  • Targeting Processive Transcription Elongation via SEC Disruption for MYC-Induced Cancer Therapy.
Targeting Processive Transcription Elongation via SEC Disruption for MYC-Induced Cancer Therapy.
Cell (2018-10-20)
Kaiwei Liang, Edwin R Smith, Yuki Aoi, Kristen L Stoltz, Hiroaki Katagi, Ashley R Woodfin, Emily J Rendleman, Stacy A Marshall, David C Murray, Lu Wang, Patrick A Ozark, Rama K Mishra, Rintaro Hashizume, Gary E Schiltz, Ali Shilatifard
ABSTRACT
The super elongation complex (SEC) is required for robust and productive transcription through release of RNA polymerase II (Pol II) with its P-TEFb module and promoting transcriptional processivity with its ELL2 subunit. Malfunction of SEC contributes to multiple human diseases including cancer. Here, we identify peptidomimetic lead compounds, KL-1 and its structural homolog KL-2, which disrupt the interaction between the SEC scaffolding protein AFF4 and P-TEFb, resulting in impaired release of Pol II from promoter-proximal pause sites and a reduced average rate of processive transcription elongation. SEC is required for induction of heat-shock genes and treating cells with KL-1 and KL-2 attenuates the heat-shock response from Drosophila to human. SEC inhibition downregulates MYC and MYC-dependent transcriptional programs in mammalian cells and delays tumor progression in a mouse xenograft model of MYC-driven cancer, indicating that small-molecule disruptors of SEC could be used for targeted therapy of MYC-induced cancer.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-SPT5 Antibody, clone 6F1, clone 6F1, from rat
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
KL-2, ≥98% (HPLC)