Skip to Content
Merck
All Photos(1)

Key Documents

1.46314

Millipore

Fluid Thioglycollate Medium

bottle capacity 250 mL, bottle filling volume 180 mL, closure type, Blue screw cap with 3 loci, pack of 20 bottles Single packed

Synonym(s):

FTM, Fluid Thioglycollate Medium, Thioglycollate Broth w. LTH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

sterility

sterile; autoclaved

Quality Level

form

liquid

feature

closure type Blue screw cap with 3 loci
ready-to-use

packaging

pack of 20 bottles Single packed

bottle capacity

250 mL

bottle filling volume

180 mL

capacity

250 mL

pH

7.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

components

histidine
tween
lecithin

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

Application

Sterility testing

Packaging

180 ml in 250 ml bottle with blue screw cap and 3 loci (6 bottles per box)

Storage and Stability

Store at +2°C to +25°C.

Other Notes

Customized product – please contact your Sales representative for further information on purchase conditions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service