1.08190
Thioglycollate broth
for microbiology
Synonym(s):
thioglycolate medium, thioglycolic medium, TG medium,
About This Item
Quality Level
form
medium granules (dehydrated (DCM))
shelf life
limited shelf life, expiry date on the label
packaging
pkg of 5 kg
pkg of 500 g
manufacturer/tradename
GranuCult®
technique(s)
microbiological culture: suitable
pH
6.9-7.3 (37 °C, 29 g/L in H2O, after autoclaving)
solubility
29 g/L
bulk density
610 kg/m3
application(s)
microbiology
pharmaceutical
storage temp.
15-25°C
suitability
anaerobic bacteria
General description
Application
Features and Benefits
- Safer - Significant reduction of inhalation of toxic and allergenic substances resulting in fewer allergic responses. Less contamination of working tools and environment
- Accurate - No separation of components and no clumping, even under humid or warm conditions, ensure a uniform product with higher performance accuracy
- Fast - Rapid and uniform dissolution in water
- Easier - Easier handling and weighing due to the better flow properties. Media do not stick to the flask or container
- Reliable - Homogenous distribution of ingredients guarantees high reproducibility even if only small amounts of media are used.
Linkage
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service