Skip to Content
Merck
All Photos(1)

Key Documents

126593

Sigma-Aldrich

Albumin, Bovine Serum, Fraction V, RIA and ELISA Grade

Synonym(s):

Albumin, Bovine Serum, Fraction V, RIA and ELISA Grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

description

Merck USA index - 14, 8468

Assay

≥98% (agarose gel electrophoresis)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤3.0 ash
≤5.0% moisture

color

off-white

solubility

0.15 M sodium chloride: 10 mg/mL

cation traces

heavy metals: ≤10 ppm

shipped in

ambient

storage temp.

2-8°C

General description

Albumin, Bovine Serum, Fraction V, RIA and ELISA Grade is a purified plasma protein that is useful as a stabilizing agent and as a carrier protein. It is prepared under non-denaturing conditions by modification of the Cohn procedure and caprylic acid heat-shock treatment.
Bovine serum albumin (BSA) is an α-helical, non-glycosylated globular protein with 17-disulfide bonds.

Application

Albumin, Bovine Serum, Fraction V, RIA and ELISA Grade has been used:
  • to block tissue sections of placenta in immunohistochemical (IHC) analysis
  • as a component of Krebs buffer for in vitro glucose-stimulated insulin secretion (GSIS) experiments and drug/palmitate preparation
  • as a vehicle control to incubate whole mouse blood to study plate aggregation in vivo

Biochem/physiol Actions

Bovine Serum Albumin (BSA), an important component in the cell culture media can transport fatty acids, hormones, and drugs. It is generally used as a blocking agent in enzyme-linked immunosorbent assay (ELISA).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine Paschalidou et al.
The Biochemical journal, 382(Pt 3), 1031-1038 (2004-07-06)
The development of renin inhibitors for the treatment of hypertension requires highly sensitive substrates to evaluate potency and to characterize the mechanism of tight-binding inhibitors. A series of intramolecularly quenched fluorogenic renin substrates, based on the N-terminal tetradecapeptide sequence of
Qi Da et al.
Microcirculation (New York, N.Y. : 1994), 25(6), e12457-e12457 (2018-04-28)
Monitoring endogenous platelets during intravital microscopy often involves two approaches: fluorescently labeled antibodies or genetic models of platelet-specific fluorescent protein expression. Due to limited data available on platelet functional changes induced by these methods, we compared functional effects of these
Lucas A J Ahrens et al.
PloS one, 12(4), e0175095-e0175095 (2017-04-06)
Controlled release drug delivery systems are well established as oral and implantable dosage forms. However, the controlled release paradigm can also be used to present complex soluble signals responsible for cellular organization during development. Endochondral ossification (EO), the developmental process
Jessica Suagee-Bedore et al.
Animals : an open access journal from MDPI, 11(12) (2021-12-25)
Older horses and those prone to obesity may be at a higher risk for inflammation than younger and leaner counterparts. Previous research indicated a postprandial elevation in plasma concentrations of interleukin-1β (IL-1β), a pro-inflammatory cytokine, after consuming 1.2 g of
Ernest T Parker et al.
Bio-protocol, 11(20), e4195-e4195 (2021-11-12)
Diffusion is a fundamental process in biological systems that governs the molecular collisions driving biochemical reactions and membrane and transport. Measurement of the diffusion coefficient and application of the Stokes-Einstein equation produces the hydrodynamic radius, which is a commonly used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service