Skip to Content
Merck
All Photos(2)

Key Documents

1.17048

Supelco

Sulfuric acid

95-97% for COD determination

Synonym(s):

Sulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
41116116
EC Index Number:
231-639-5
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Sulfuric acid 95 - 97% for COD determination,

vapor density

<0.3 (25 °C, vs air)

Quality Level

vapor pressure

0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)

form

liquid

analyte functional class(es)

chemical oxygen demand (COD)

concentration

≤5 mg/L (CSB)

technique(s)

photometry: suitable

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

density

1.840 g/mL at 25 °C (lit.)

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

storage temp.

15-25°C

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer this sulfuric acid for the measurement of COD in seawater and other applications to be purchased together with the tests. When measuring COD e.g. in seawater with Spectroquant® COD Cell Test Kits 1.17058, 1.17059 one needs to measure a blank as reference. More information can be found in the instruction leaflet.
The COD (chemical oxygen demand) expresses the amount of oxygen originating from potassium dichromate that reacts with the oxidizable substances contained in 1 l of water under the working conditions of the specified procedure.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service