Skip to Content
Merck
All Photos(2)

Key Documents

1.12080

Supelco

Sulfuric acid

98%, for analysis EMSURE®

Synonym(s):

Sulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-639-5
NACRES:
NA.21

Product Name

Sulfuric acid 98%, for analysis EMSURE®

vapor density

<0.3 (25 °C, vs air)

Quality Level

vapor pressure

0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)

product line

EMSURE®

Assay

≥98% (alkalimetric)

form

liquid

impurities

≤0.0002% Substances reducing potassium permanganate (as SO2)

ign. residue

≤0.0005%

color

APHA: ≤10

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.00001%
nitrate (NO3-): ≤0.00002%
phosphate (PO43-): ≤0.00005%

cation traces

Ag: ≤0.000002%
Al: ≤0.000005%
As: ≤0.000001%
Ba: ≤0.000005%
Be: ≤0.000001%
Bi: ≤0.00001%
Ca: ≤0.00002%
Cd: ≤0.000002%
Co: ≤0.000001%
Cr: ≤0.000005%
Cu: ≤0.000001%
Fe: ≤0.00001%
Ge: ≤0.000005%
K: ≤0.00001%
Li: ≤0.000001%
Mg: ≤0.000005%
Mn: ≤0.000001%
Mo: ≤0.000005%
NH4+: ≤0.0002%
Na: ≤0.00005%
Ni: ≤0.000002%
Pb: ≤0.000002%
Se: ≤0.0001%
Sr: ≤0.000002%
Ti: ≤0.00001%
Tl: ≤0.000005%
V: ≤0.000001%
Zn: ≤0.000005%
Zr: ≤0.00001%

SMILES string

OS(O)(=O)=O

storage temp.

no temp limit

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Simultaneous determination of monochloropropanediol esters and glycidyl esters in vegetable oils by acidic transesterification-gas chromatography-mass spectrometry.: This study by Wang et al. presents a method for detecting harmful compounds in vegetable oils using sulfuric acid for transesterification, followed by GC-MS analysis (Wang et al., 2022).


  • Evaluation of an improved acid hydrolysis-HPLC assay for the acetyl content in chitin and chitosan.: Ng et al. developed an enhanced HPLC method for measuring acetyl content in biopolymers, utilizing sulfuric acid for hydrolysis to ensure accurate quantification (Ng et al., 2006).

Analysis Note

Assay (alkalimetric): ≥ 98.0 %
Colour: ≤ 10 Hazen
Chloride (Cl): ≤ 0.00001 %
Nitrate (NO₃): ≤ 0.00002 %
Phosphate (PO₄): ≤ 0.00005 %
Ag (Silver): ≤ 0.000002 %
Al (Aluminium): ≤ 0.000005 %
As (Arsenic): ≤ 0.000001 %
Ba (Barium): ≤ 0.000005 %
Be (Beryllium): ≤ 0.000001 %
Bi (Bismuth): ≤ 0.00001
Ca (Calcium): ≤ 0.00002 %
Cd (Cadmium): ≤ 0.000002 %
Co (Cobalt): ≤ 0.000001 %
Cr (Chromium): ≤ 0.000005 %
Cu (Copper): ≤ 0.000001 %
Fe (Iron): ≤ 0.00001
Ge (Germanium): ≤ 0.000005 %
K (Potassium): ≤ 0.00001 %
Li (Lithium): ≤ 0.000001 %
Mg (Magnesium): ≤ 0.000005 %
Mn (Manganese): ≤ 0.000001 %
Mo (Molybdenum): ≤ 0.000005 %
NH₄ (Ammonium): ≤ 0.0002 %
Na (Sodium): ≤ 0.00005 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.000002 %
Se (Selenium): ≤ 0.0001 %
Sr (Strontium): ≤ 0.000002 %
Ti (Titanium): ≤ 0.00001
Tl (Thallium): ≤ 0.000005 %
V (Vanadium): ≤ 0.000001 %
Zn (Zinc): ≤ 0.000005 %
Zr (Zirconium): ≤ 0.00001
Substances reducing potassium permanganate (as SO₂): ≤ 0.0002 %
Residue on ignition: ≤ 0.0005 %
Date of expiry: see product label

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Glass bottles are ideal for storing acids because they are inert and offer a long shelf life.

Glass bottles are ideal for storing acids because they are inert and offer a long shelf life.

Glass bottles are ideal for storing acids because they are inert and offer a long shelf life.

Glass bottles are ideal for storing acids because they are inert and offer a long shelf life.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service