Z557463
Julabo® bath spring clamps for Erlenmeyer flasks
fits 500 mL flasks
All Photos(1)
About This Item
UNSPSC Code:
41122413
NACRES:
NB.77
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970608
General description
The individual spring clips can be positioned in the shaking water bath using the base tray for spring clips (Z743201-1EA). A maximum of 6 spring clips for 500 ml Erlenmeyer flasks can be mounted on a base tray.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.