Key Documents
Z151491
Sigma-Aldrich
Bransonic® ultrasonic cleaner accessories
Solid tray, stainless steel, tank capacity 5.7 L
About This Item
UNSPSC Code:
41101500
NACRES:
NB.74
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Branson 100-410-174
tank capacity
5.7 L
Related Categories
General description
Solid Stainless steel tray for use with Branson M series, model 3800 Utrasonic Bath. Dimensions 220 mm x 110 mm x 100 mm.
Legal Information
Bransonic is a registered trademark of Branson Ultrasonics Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)