Documents
Z305359
Sigma-Aldrich
Bransonic® Model B200 cleaner
AC/DC input 240 V AC
Synonym(s):
ultrasonic bath sonicator, ultrasonic cleaning bath
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
feature
CE compliant
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Branson 100-951-011 (NA)000-001-100 (Europe)
General description
Low-cost, portable, ultrasonic cleaner has the same cleaning ability as larger models. SS tank with Noryl® base, parts basket, and plastic cover. Cleaner automatically shuts off after 5 min.
Legal Information
Bransonic is a registered trademark of Branson Ultrasonics Corp.
Noryl is a registered trademark of Sabic Innovative Plastics IP B.V.