Key Documents
Z117951
Luer-to-Closed end stopper
male Luer lock to closed end
About This Item
UNSPSC Code:
41122500
NACRES:
NB.03
manufacturer/tradename
Cadence Science
Micro-Mate
General description
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes.
316 SS fittings are noncontaminating, easy to clean and resistant to corrosion. Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Choose from one of the most recent versions:
Certificates of Analysis (COA)