CLS210325
Pyrex® class A colored scale burette, with PTFE stopcock plug
volume 25 mL, accuracy: 0.03 mL
Synonym(s):
burettes, glass burettes, Corning® 2103
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
Recommended Products
material
borosilicate glass
feature
subdivision 0.1 mL
manufacturer/tradename
Corning 2103-25
volume measuring range
25 mL, accuracy: 0.03 mL
volume
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Has markings which are permanently fused into glass; fine, sharp lines; easy-to-read numbers. Smooth, microfinish stopcock barrel assures leak resistance. Has 2mm bore PTFE stopcock plug and dustcover.
Other Notes
Class A burettes—Tolerances established by ASTM E 694 for volumetric ware, ASTM E 542 for calibration of volumetric ware, and ASTM E 287 for burettes.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service