688827
trans-1-Phenyl-2-buten-1-one
technical, 80%
Synonym(s):
trans-Crotonophenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H10O
CAS Number:
Molecular Weight:
146.19
Beilstein:
2039624
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical
Assay
≥80% (GC)
80%
form
liquid
functional group
ketone
phenyl
storage temp.
2-8°C
SMILES string
C\C=C\C(=O)c1ccccc1
InChI
1S/C10H10O/c1-2-6-10(11)9-7-4-3-5-8-9/h2-8H,1H3/b6-2+
InChI key
FUJZJBCWPIOHHN-QHHAFSJGSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service