Skip to Content
Merck
All Photos(1)

Key Documents

306940

Sigma-Aldrich

2,6-Dimethylbenzenethiol

95%

Synonym(s):

2,6-Dimethylthiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3SH
CAS Number:
Molecular Weight:
138.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.5749 (lit.)

bp

122 °C/50 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(C)c1S

InChI

1S/C8H10S/c1-6-4-3-5-7(2)8(6)9/h3-5,9H,1-2H3

InChI key

QCLJODDRBGKIRW-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Dimethylbenzenethiol was used in the synthesis of (2,6-Me2C6H3S)2Pb by reacting with Pb(OAc)2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah E Appleton et al.
Dalton transactions (Cambridge, England : 2003), (21)(21), 3515-3520 (2004-10-29)
The amine coordination of lead(II) has been examined through the preparation and structural analysis of Lewis base adducts of bis(thiolato)lead(II) complexes. Reaction of Pb(OAc)(2) with 2,6-dimethylbenzenethiol affords (2,6-Me(2)C(6)H(3)S)(2)Pb (6) in high yield. The solubility of 6 in organic solvents allows

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service