222348
Diacetone acrylamide
contains ≤100 ppm inhibitor, 99%
Synonym(s):
N-(1,1-Dimethyl-3-oxobutyl)acrylamide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
CH2=CHCONHC(CH3)2CH2COCH3
CAS Number:
Molecular Weight:
169.22
Beilstein:
1928444
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99%
contains
≤100 ppm inhibitor
mp
54-56 °C (lit.)
SMILES string
CC(=O)CC(C)(C)NC(=O)C=C
InChI
1S/C9H15NO2/c1-5-8(12)10-9(3,4)6-7(2)11/h5H,1,6H2,2-4H3,(H,10,12)
InChI key
OMNKZBIFPJNNIO-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Latex films with gradients in crosslink density created by small-molecule-based auto-stratification.
Heike Römermann et al.
The European physical journal. E, Soft matter, 42(2), 21-21 (2019-02-23)
A suitable balance of convective and diffusive transport of small molecules contained in the liquid phase of a drying latex film leads to auto-stratification and to functionally graded films. Differing from blends of latex particles, which may also experience drying-induced
Aki Ikeda et al.
AMB Express, 7(1), 101-101 (2017-05-27)
Under bioassay-guided investigation, a sporulation-inducing factor (SIF) toward Bacillus spp. was searched for in methanol (MeOH) extracts of soybean curd residues, and diacetonamine (1) was identified as the active compound. SIF was first isolated as a monoacetylated derivative (2, 4.1 mg
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service