Skip to Content
Merck
All Photos(1)

Key Documents

1605523

USP

Ropivacaine Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(R)-Ropivacaine hydrochloride monohydrate, (2R)-N-(2,6-Dimethylphenyl)-1-propyl-2-piperidinecarboxamide hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26N2O · HCl · H20
CAS Number:
Molecular Weight:
331.02
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ropivacaine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C17H26N2O.ClH/c1-4-11-19-12-6-5-10-15(19)17(20)18-16-13(2)8-7-9-14(16)3;/h7-9,15H,4-6,10-12H2,1-3H3,(H,18,20);1H/t15-;/m1./s1

InChI key

NDNSIBYYUOEUSV-XFULWGLBSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ropivacaine Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fentanyl Citrate and Ropivacaine Hydrochloride Compounded Injection
  • Ropivacaine Hydrochloride
  • Ropivacaine Hydrochloride Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Heart

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service