Skip to Content
Merck
All Photos(1)

Key Documents

42123

Supelco

Mettler-Toledo Calibration substance ME 30130599, Sodium acetate anhydrous

tracable to primary standards (LGC)

Synonym(s):

Sodium acetate, Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

form

solid

autoignition temp.

1112 °F

shelf life

limited shelf life, expiry date on the label

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium acetate is the sodium salt of acetic acid with a wide variety of chemical, biochemical and food applications of both laboratory and industrial scale.

Mettler-Toledo calibration substance ME 30130599, sodium acetate anhydrous is an analytical standard, ideal for the regular monitoring of Mettler-Toledo melting point instrument. Its value is a mean of 6 to 12 iterations with a Mettler-Toledo MP90 Excellence instrument that is calibrated against primary standards. The melting point is validated by Capillary method according to European Pharmacopeia (2.2.14.)

Application

Sodium acetate anhydrous Mettler-Toledo calibration substance ME 30130599 is used to calibrate Mettler-Toledo melting point instruments.

Features and Benefits

  • Melting point calibration standard traceable to a primary standard (LGC)
  • Melting point measured in both thermodynamic and pharmacopeia modes for physically correct and heating rate dependent melting point analyses, respectively
  • Available with certificates of analysis and safety data sheet
  • A product of analytical standard grade to help meet the QC/QA requirements of melting point determination
  • Measurement of uncertainty up to ± 0.4 °C

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service