Skip to Content
Merck
All Photos(1)

Key Documents

15809

Supelco

Mettler-Toledo Calibration substance ME 30130597, p-Toluamide

traceable to primary standards (LGC)

Synonym(s):

p-Toluamide, 4-Methylbenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CONH2
CAS Number:
Molecular Weight:
135.16
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

shelf life

limited shelf life, expiry date on the label

mp

161-163 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

Cc1ccc(cc1)C(N)=O

InChI

1S/C8H9NO/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3,(H2,9,10)

InChI key

UHBGYFCCKRAEHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mettler-Toledo calibration substance ME 30130597, p-toluamide is an analytical standard, ideal for use in the routine calibration of Mettler-Toledo melting point instrument. Its value is measured as a mean of 6 to 12 iterations using a Mettler-Toledo MP90 Excellence instrument that is calibrated with primary standards. The melting point determination is carried out by Capillary method in accordance with European Pharmacopeia (2.2.14.)

Application

p-Toluamide Mettler-Toledo calibration substance ME 30130597 is used to calibrate Mettler-Toledo melting point instruments.

Features and Benefits

  • Melting point calibration standard traceable to a primary standard (LGC, London)
  • Melting point evaluated in both thermodynamic and pharmacopeia modes
  • Provided with certificates of analysis and safety data sheet
  • A product of analytical standard grade to help meet the QC/QA requirements of melting point determination

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service