GF418-MG
IL-1 beta, Human
HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
HumanKine growth factors are expressed in a proprietary human 293 protein expression system and are manufactured without the use of any animal products and are considered Xeno-Free.
Interleukin-1 (IL-1) is a cytokine that is thought to be connected to neurodegeneration and in central nervous system-mediated defense reactions to inflammation. Interleukin-1 beta (IL-1B) maintains mitogen-induced proliferation, IL-2 production and expression of IL-2R. IL-1 beta also seems to have the ability to boost the number of cells that move into the G1 phase of the cell cycle. IL-1 beta also has the capacity to stimulate apoptosis on normal human fetal membranes. IL-1 beta may play an important role in inflammation regulating cytokine networks.
Quality
Bioactivity assay: Determined by the dose-dependent stimulation of the proliferation of mouse D10S cells. ED50 is typically 0.02-0.08 ng/ml.
HumanKine IL-1 beta is manufactured using all non-animal reagents and is considered Xeno-Free
Physical form
Product is presented in PBS and filtered through a 0.2 micron filter before lyophilization.
Storage and Stability
Store at -20°C for up to 4 months from date of receipt. Further dilutions should be made into appropriate buffer or medium to which protein has been added.
Legal Information
HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service