Skip to Content
Merck
All Photos(1)

Key Documents

407615-M

Sigma-Aldrich

Interleukin-1β, Human, Recombinant, E. coli

Synonym(s):

Interleukin-1β, Human, Recombinant, E. coli, rhIL-1β

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202

Assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0 EU/μg Endotoxin (EU/μg IL-1β)

storage temp.

−70°C

General description

Recombinant, human interleukin-1β expressed in E. coli. Major form of interleukin-1 secreted by monocytes and macrophages. IL-1β is activated by proteolytic cleavage of the precursor molecule. Increases nitric oxide production in smooth muscle cells and suppresses apoptosis in rat ovarian follicles. Involved in inflammatory and immune responses and promotes wound healing. Induces nitric oxide synthase in insulin-producing cells. 1 µg of IL-1β corresponds to 1.9 x 105 WHO units.
Recombinant, human interleukin-1β expressed in E. coli. Major form of interleukin-1 secreted by monocytes and macrophages. Induces nitric oxide synthase in pancreatic and smooth muscle cells and suppresses apoptosis in rat ovarian follicles. Involved in inflammatory and immune responses and promotes wound healing.

Biochem/physiol Actions

ED₅₀ = <12 pg/ml as measured in a cell proliferation assay using the murine T-cell line, D10.G4.1 (Symons, J.A. et al. 1987)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered PBS, 50 µg BSA/µg IL-1β.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C) for long term storage or refrigerate (4°C) for short term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -70°C.
Reconstitute to a concentration of ≥10 µg/ml with sterile PBS containing ≥0.1% HSA or BSA.

Other Notes

Chun, S.Y., et al. 1995. Endocrinology136, 3120.
Kunz, D., et al. 1994. Biochem. J. 304, 337.
Eizirik, D.L., et al. 1993. FEBS Lett. 317, 62.
Dinarello, C>A> 1991. Blood77, 1627.
Casagli, M.C., et al. 1989. Biochem. Biophys. Res. Commun.162, 357.
Symons, J.A., et al. 1987. In Lymphokines and interferons, A Practical Approach (Clemens, M.J., et al. eds.) IRL Press, Oxford p. 272.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service