Skip to Content
Merck
All Photos(1)

Key Documents

71341

Sigma-Aldrich

pRSFDuet-1 DNA - Novagen

Synonym(s):

pRSFDuet-1 Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106602
NACRES:
NA.51

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

Promoter

Promoter name: T7

selection

kanamycin

shipped in

wet ice

storage temp.

−70°C

General description

pRSFDuet-1 is designed for the co-expression of two target proteins. The vector encodes two multiple cloning sites (MCS) each of which is preceded by a T7 promoter, lac operator, and ribosome binding site (RBS). The vector also carries the pRSF1030 replicon (also known as NTP1), LacI gene, and kanamycin resistance marker.

Application

pRSFDuet-1 DNA - Novagen has been used as a vector:
  • to clone the DNA fragments of defensin α 5 (DEFA5) for recombinant expression
  • to clone the coding region of Mad1597–719
  • to clone the globular domain of Bombyx mori Spc24 (73-162aa) with an N-terminal 6X His-tag to generate the B. mori 6xHis-Spc24(73-162)-Spc25(70-211) pRSF-DUET1 bacterial expression constructs

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiao Qiao et al.
Experimental cell research, 407(2), 112809-112809 (2021-09-07)
Defensins are highly conserved antimicrobial peptides, which ubiquitously expressed in different species. In addition to the functions in host defense, their aberrant expression have also been documented in cancerous tissue including breast cancer, lung caner and renal carcinoma etc. Whereas
Elyse S Fischer et al.
EMBO reports, 22(7), e52242-e52242 (2021-05-21)
During metaphase, in response to improper kinetochore-microtubule attachments, the spindle assembly checkpoint (SAC) activates the mitotic checkpoint complex (MCC), an inhibitor of the anaphase-promoting complex/cyclosome (APC/C). This process is orchestrated by the kinase Mps1, which initiates the assembly of the
Mohannad Al Chiblak et al.
BMC molecular and cell biology, 20(1), 60-60 (2019-12-21)
ZNF746 and ZNF777 belong to a subset of the large Krüppel-associated box (KRAB) zinc finger (ZNF) transcription factor family. They contain, like four other members in human, an additional conserved domain, the "domain of unknown function 3669" (DUF3669). Previous work
Nuria Cortes-Silva et al.
Current biology : CB, 30(4), 561-572 (2020-02-08)
Accurate chromosome segregation requires assembly of the multiprotein kinetochore complex at centromeres. In most eukaryotes, kinetochore assembly is primed by the histone H3 variant CenH3 (also called CENP-A), which physically interacts with components of the inner kinetochore constitutive centromere-associated network

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service