Skip to Content
Merck
All Photos(1)

Key Documents

5.30650

Sigma-Aldrich

Luciferase Substrate, CycLuc1

Synonym(s):

Luciferase Substrate, CycLuc1, 2-(6,7-dihydro-5h-thiazolo[4,5-f]indol-2-yl)-4,5-dihydrothiazole-4-carboxylicacid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3O2S2
Molecular Weight:
305.38
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

Assay

≥94% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A synthetic cyclic alkylamino luciferin that exhibitsproperties that are superior to those of D-luciferin. Binds to luciferase with higheraffinity and emits about 5.7-fold more light than aminoluciferin and3.2-fold more light than D-luciferin. Its superior light output is maintainedover a wide range (1-100 µM). Shown to readily cross the blood-brain barrier inmice and provides much improved noninvasive bioluminescence imaging (BLI) ofthe brain at a lower dose when compared to D-luciferin. Yields about 10-foldhigher bioluminescent signal than D-luciferin when Luc2 luciferase expressing4T1 breast cancer cells are implanted into the mammary fat pads of BABL/c mice.A rapid (4-5 min), long-lasting, and steady signal peak is observed followingan i.v injection of CycLuc1 in mice.

Application

Luciferase Substrate, CycLuc1 has been used for transfecting HEK293 cells to study the inhibition of intracellular luciferase crystallization by D-luciferin and its synthetic analogues.[1]

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Luciferase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haruki Hasegawa
Experimental cell research, 440(1), 114131-114131 (2024-06-15)
Firefly luciferase (Fluc) from Photinus pyralis is one of the most widely used reporter proteins in biomedical research. Despite its widespread use, Fluc's protein phase transition behaviors and phase separation characteristics have not received much attention. Current research uncovers Fluc's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service