Key Documents
CLS9999241
Corning® phenolic cap with white rubber liner
24-410 thread
Synonym(s):
phenolic caps, screw caps
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
manufacturer/tradename
Corning 9999-241
fitting
24-410 thread
General description
These reusable Corning caps are fabricated from specially formulated phenolic resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in white rubber liner.
Legal Information
Corning is a registered trademark of Corning, Inc.