Key Documents
CLS999824
Corning® phenolic cap, PTFE liner
24-400 thread
Synonym(s):
Corning® 9998
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
PTFE liner
black phenolic cap
manufacturer/tradename
Corning 9998-24
H
~12.5 mm
fitting
24-400 thread
General description
These reusable Corning caps are fabricated from specially formulated resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface to provide a highly inert sealing face.
Legal Information
Corning is a registered trademark of Corning, Inc.