BR781345
BRAND® 384-well PCR plate
full skirt, clear, cut corner A12, H12
Synonym(s):
PCR, PCR Plates, Plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
material:
clear
polypropylene
polypropylene
size:
384 well
sterility:
non-sterile
binding type:
non-treated surface
feature:
skirt
full skirt
full skirt
Recommended Products
material
clear
polypropylene
sterility
non-sterile
feature
skirt
full skirt
packaging
pack of 50 ea
manufacturer/tradename
BRAND 781345
technique(s)
PCR: suitable
size
384 well
working volume
30 μm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
The 40 μL wells can hold sample volumes of 2 μL to 30 μL. This helps to reduce reagents costs and shortens cycle times. The plates can be filled using multichannel pipettes or robotic systems. The ultra thin-walled PCR plate design facilitates constant, rapid and precise heat transfer. The smooth vessel interior minimizes the binding of enzymes and nucleic acid to the walls. The raised lips of the wells protect against cross-contamination, allowing a reliable seal with the sealing mat tailored to the plates.
Features and Benefits
- For applications in automation and high-throughput analyses
- Rapid heat transfer through thin vessel walls
- Can be sealed reliably with self-adhesive sealing films
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service