Skip to Content
Merck
All Photos(1)

Key Documents

CLS3702

Corning® 384 well microplate

clear polystyrene, flat bottom, non-sterile, lid: no, case of 100

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
clear bottom
clear polystyrene
flat bottom
size:
384 wells
Pricing and availability is not currently available.

material

clear bottom
clear polystyrene
flat bottom

sterility

non-sterile

feature

lid: no
skirt

packaging

case of 100

manufacturer/tradename

Corning 3702

size

384 wells

well volume

112 μL

well working volume

20-80 μL

wells

384 wells

color

clear

suitability

suitable for (cell cuture)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

For colorimetric assays

Features and Benefits

  • Corning® 384 well microplates support various application requirements.
  • Non-treated clear polystyrene surface is suitable primarily for the immobilization of large molecules, such as antibodies.
  • The flat bottom is usually the best choice for detection.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service