Skip to Content
Merck
All Photos(1)

Key Documents

254363

Sigma-Aldrich

Methylmagnesium iodide solution

3.0 M in diethyl ether

Synonym(s):

Iodomethylmagnesium, Methylmagnesium iodine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3MgI
CAS Number:
Molecular Weight:
166.24
Beilstein:
1209226
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

3.0 M in diethyl ether

density

1.261 g/mL at 25 °C

SMILES string

C[Mg]I

InChI

1S/CH3.HI.Mg/h1H3;1H;/q;;+1/p-1

InChI key

AUPXBVDHVRZMIB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Methylmagnesium iodide solution can be used:
  • To prepare cis-1,2-dialkylcyclopropanols by treating with titanium(IV) alkoxide and carboxylic esters.[1]
  • In stereospecific Ni catalyzed Kumada cross-coupling reactions of benzylic ethers.[2]
  • To prepare 2,4-disubstituted selenochromenes by reacting with 1-benzoselenopyrylium salts.[3]
  • In one of the key synthetic steps for the preparation of 7β-methyl-substituted 5-androstene derivatives from 3β-acetoxyandrost-5-en-17-one stereoselectively.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel stereoselective synthesis of 7β -methyl-substituted 5-androstene derivatives
Zheng Y and Li Y
The Journal of Organic Chemistry, 68(4), 1603-1606 (2003)
Advanced Procedure for the Preparation of cis-1, 2-Dialkylcyclopropanols-Modified Ate Complex Mechanism for Titanium-Mediated Cyclopropanation of Carboxylic Esters with Grignard Reagents
Kulinkovich OG and Kananovich DG
European Journal of Organic Chemistry, 2007(13), 2121-2132 (2007)
Reaction of 1-benzoselenopyrylium salts with grignard reagents: Formation of 2, 4-disubstituted selenochromenes and their conversion into the corresponding 1-benzoselenopyrylium salts
Sashida H, et al.
Journal of Heterocyclic Chemistry, 39(2), 405-409 (2002)
Stereospecific nickel-catalyzed cross-coupling reactions of benzylic ethers with isotopically-labeled grignard reagents
Dawson DD and Jarvo ER
Organic Process Research & Development, 19(10), 1356-1359 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service