Skip to Content
Merck
All Photos(2)

Key Documents

189901

Sigma-Aldrich

Methylmagnesium chloride solution

3.0 M in THF

Synonym(s):

Chloromethylmagnesium, Methyl magnesium chloride, Methylmagnesiumchloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3MgCl
CAS Number:
Molecular Weight:
74.79
Beilstein:
3587199
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

autoignition temp.

609 °F

reaction suitability

reaction type: Grignard Reaction

concentration

3.0 M in THF

density

1.013 g/mL at 25 °C

SMILES string

C[Mg]Cl

InChI

1S/CH3.ClH.Mg/h1H3;1H;/q;;+1/p-1

InChI key

RQNMYNYHBQQZSP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Methylmagnesium chloride solution (3 M in THF) undergoes 1,3-addition with dialdose derived nitrones to form N-benzylhydroxylamines. The reductive cleavage of the N-O bond of these N-benzylhydroxylamines results in N-benzylaminosugars, which are key moieties of bioactive compounds. It can also react with Garner type aldehydes to form the corresponding secondary alcohols.[1]

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new route to aminosugars from sugar nitrones: synthesis of 6-deoxynojirimycin.
Dhavale DD
Tetrahedron Asymmetry, 8(9), 1475-1486 (1997)

Articles

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Questions

1–7 of 7 Questions  
  1. Should I store the product under inert conditions?

    1 answer
    1. This product is packaged in Sure/Seal bottles and closures in order to preserve the quality of the reagent. Exposure to air and light may result in degradation. To maintain the shelf life and quality, larger package sizes may be stored in aliquots using smaller Sure/Seal bottles and Crown caps. Please see the link below to review these product options:
      Crown Caps - for Sure/Seal bottles
      https://www.sigmaaldrich.com/product/aldrich/z102148
      Sure/Seal bottles
      https://www.sigmaaldrich.com/substance/suresealbottles1234598765

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. In one litere of tetra hydra furan THF, how much of methyl megnesium chloride should be requried?

    1 answer
    1. The molar mass of methylmagnesium chloride is 74.79 g/mol. This product is a 3.0 M methylmagnesium chloride solution in THF. This corresponds to 224.37 grams of methylmagnesium chloride per liter of THF to achieve this concentration.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. Is the BHT inhibitor present in Product 189901, Methylmagnesium chloride solution?

    1 answer
    1. Yes, BHT is present in the THF. This information can be found on the MSDS, which is available from our website.

      Helpful?

  7. Can Product 189901, Methylmagnesium chloride solution, be used if a precipitate is present?

    1 answer
    1. If a precipitate is present, the product can be heated and stirred until the precipitate is dissolved.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service