Merck
HomeGas Chromatography (GC) Water Determination in Liquefied Petroleum Gas using GC BID and Ionic Liquid Column Watercol™
Water Determination in Liquefied Petroleum Gas using GC BID and Ionic Liquid Column Watercol™
Shimadzu Application Data Sheet No.18
Water in petrochemical feedstocks can cause problems for processors. Freezing of pipe lines and valves and poisoning of expensive catalysts are just a few examples.
Monitoring water in petroleum from an upstream source to the downstream processing plant is critical to insure uninterrupted operation.
A new determination option by GC can bypass undesirable chemical interference effects from the petroleum to the “Sulfur interaction” that can skew results obtained by traditional water determination techniques. Shimadzu’s proprietary Barrier Ionization Discharge (BID) detector and Supelco’s water analysis column “Watercol” are combined to separate and measure the water in a formulation of feedstock (Figure 1) and provide a sensitive and accurate result (Table 1). Measurements can be made down to sub-ppm level of water detection.
Chromatogram for water determination (25 ppm) in LPG. Quantification of Limit(S/N=10) and Detection of Limit(S/N=3.3) can be down to 0.66 ppm and 0.22 ppm respectively
Figure 1.Chromatogram for water determination (25 ppm) in LPG. Quantification of Limit(S/N=10) and Detection of Limit(S/N=3.3) can be down to 0.66 ppm and 0.22 ppm respectively
Instrument
Gas Chromatograph:Tracera (GC-2010 Plus A + BID-2010 Plus)
Sample Injection:Valco Internal Liquid Sample Injector with Splitter Injection Unit
Gas Purifier:Supelco High Capacity Gas Purifier (Cat.#29541-U)
Analysis Conditions
Column:Watercol™ 1910, 60 m x 0.25 mm ID, 0.20 μm
Oven:35 °C (2.0 min) – 5 °C/min – 150 °C (15 min) Total. 40 min
Carrier Gas:Helium 45 cm/sec (Column flow rate 3.78 mL/min)
Inj. Volume:2 μL
Split:1:5
Transfer Line Temp.:175 °C (After Internal Liquid Sample Injector to GC column Oven)
WaterNo.1No.2No.3No.4No.5AverageStandard Deviation%RSD
RT(min)14.28514.28814.28614.29614.30414.2920.0080.06
Area(mV•s)244,037249,854246,884242,950238,428244,4304,2961.76
Height(mV)12,41812,60012,46812,04511,85112,2763152.57
Conc.(ppm)24.9625.5625.2524.8524.3925.000.441.76
Table 1.Repeatability of Water analysis(n=5).
This application is also subject of the ASTM Work Item www.astm.org/DATABASE.CART/WORKITEMS/WK59649.htm
More information on Watercol™ Columns.
Download the Shimadzu Data Sheet.
Materials
Desculpe, ocorreu um erro inesperado
Network error: Failed to fetch